Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
SLC17A3
Entrez Gene ID
10786
Gene Name
solute carrier family 17 member 3
Gene Aliases
GOUT4, NPT4, UAQTL4
Chromosome Location
Chr.6: 25845100 - 25874243 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001098486.1 NP_001091956.1 10 - 11 1379 84
NM_006632.3 NP_006623.2 9 - 10 1145 84
GenBank mRNA AK298271.1 - 10 - 11 1379 84
BC017952.1 - 9 - 10 1114 84
CB163247.1 - 4 - 5 517 84
U90545.1 - 9 - 10 1354 84

View Full Product Details