The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_005074.3 | NP_005065.2 | 4 - 5 | 558 | 60 |
XM_011514818.2 | XP_011513120.2 | 4 - 5 | 595 | 60 | |
XM_011514819.2 | XP_011513121.2 | 4 - 5 | 595 | 60 | |
XM_011514820.2 | XP_011513122.2 | 4 - 5 | 595 | 60 | |
XM_011514821.2 | XP_011513123.1 | 2 - 3 | 312 | 60 | |
XM_017011199.1 | XP_016866688.1 | 4 - 5 | 595 | 60 | |
XM_017011200.1 | XP_016866689.1 | 4 - 5 | 595 | 60 | |
XM_017011201.1 | XP_016866690.1 | 4 - 5 | 595 | 60 | |
XM_017011202.1 | XP_016866691.1 | 3 - 4 | 523 | 60 | |
GenBank mRNA | AK290783.1 | - | 4 - 5 | 540 | 60 |
AK308035.1 | - | 4 - 5 | 540 | 60 | |
AY780791.1 | - | 3 - 4 | 442 | 60 | |
BC101745.1 | - | 3 - 4 | 495 | 60 | |
BC101747.1 | - | 3 - 4 | 495 | 60 | |
D28532.1 | - | 3 - 4 | 439 | 60 | |
X71355.1 | - | 3 - 4 | 454 | 60 |