Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CFD
Entrez Gene ID
1675
Gene Name
complement factor D
Gene Aliases
ADIPSIN, ADN, DF, PFD
Chromosome Location
Chr.19: 859659 - 863569 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001928.3 NP_001919.2 1 - 2 72
GenBank mRNA AJ313463.1 - 1 - 2 72
AK300963.1 - 1 - 2 72
BC057807.1 - 1 - 2 72
BG573629.1 - 1 - 2 72
DQ884409.1 - 1 - 2 79 93
HY087483.1 - 1 - 2 72

View Full Product Details