The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_032871.3 | NP_116260.2 | 2 - 3 | 275 | 70 |
NM_152222.1 | NP_689408.1 | 2 - 3 | 213 | 70 | |
XM_006718714.2 | XP_006718777.1 | 2 - 3 | 178 | 70 | |
XM_011545305.1 | XP_011543607.1 | 2 - 3 | 178 | 70 | |
GenBank mRNA | AF319553.1 | - | 2 - 3 | 275 | 70 |
AK027899.1 | - | 2 - 3 | 213 | 70 | |
BC017279.2 | - | 2 - 3 | 138 | 70 | |
BC051810.1 | - | 2 - 3 | 165 | 70 |