The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_032871.3 | NP_116260.2 | 9 - 10 | 1276 | 65 |
NM_152222.1 | NP_689408.1 | 9 - 10 | 1214 | 65 | |
XM_005274356.2 | XP_005274413.1 | 4 - 5 | 1377 | 65 | |
XM_006718714.2 | XP_006718777.1 | 7 - 8 | 841 | 65 | |
XM_011545305.1 | XP_011543607.1 | 8 - 9 | 1099 | 65 | |
GenBank mRNA | AF319553.1 | - | 9 - 10 | 1276 | 65 |
AK027899.1 | - | 9 - 10 | 1214 | 65 | |
AK057036.1 | - | 3 - 4 | 1018 | 65 | |
AK310103.1 | - | 6 - 7 | 1082 | 65 | |
BC017279.2 | - | 9 - 10 | 1139 | 65 | |
BC051810.1 | - | 9 - 10 | 1166 | 65 |