Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
CCDC88B
Entrez Gene ID
283234
Gene Name
coiled-coil domain containing 88B
Gene Aliases
BRLZ, CCDC88, HKRP3, gipie
Chromosome Location
Chr.11: 64340203 - 64357534 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_032251.5 NP_115627.6 20 - 21 3554 66
XM_006718519.3 XP_006718582.1 20 - 21 3566 66
XM_011544957.2 XP_011543259.1 19 - 20 3804 66
XM_011544959.2 XP_011543261.1 19 - 20 3804 66
XM_011544960.2 XP_011543262.1 19 - 20 3470 66
XM_011544962.2 XP_011543264.1 6 - 7 1426 66
GenBank mRNA AK074126.1 - 13 - 14 3208 66
AK090436.1 - 19 - 20 3767 66
AY491401.1 - 3 - 4 275 66
BC040232.1 - 7 - 8 1111 66
BC140822.1 - 20 - 21 3543 66
BC140824.1 - 20 - 21 3543 66
BC141866.1 - 20 - 21 3533 66

View Full Product Details