The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_000494.3 | NP_000485.3 | 41 - 42 | 2960 | 84 |
GenBank mRNA | AB900157.1 | - | 40 - 41 | 2791 | 84 |
M91669.1 | - | 41 - 42 | 2896 | 84 |