Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
SFXN3
Entrez Gene ID
81855
Gene Name
sideroflexin 3
Gene Aliases
BA108L7.2, SFX3
Chromosome Location
Chr.10: 101031229 - 101041243 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_030971.3 NP_112233.2 10 - 11 1287 150
GenBank mRNA AK074707.1 - 9 - 10 900 150
AK299405.1 - 9 - 10 906 150
BC000124.1 - 9 - 10 893 150

View Full Product Details