Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SFXN3
Entrez Gene ID
81855
Gene Name
sideroflexin 3
Gene Aliases
BA108L7.2, SFX3
Chromosome Location
Chr.10: 101031229 - 101041243 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_030971.3 NP_112233.2 1 - 2 291 150
GenBank mRNA AL701236.1 - 1 - 2 297 150
BI819328.1 - 1 - 2 227 151

View Full Product Details