Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SFXN3
Entrez Gene ID
81855
Gene Name
sideroflexin 3
Gene Aliases
BA108L7.2, SFX3
Chromosome Location
Chr.10: 101031229 - 101041243 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_030971.3 NP_112233.2 3 - 4 635 73
GenBank mRNA AK074707.1 - 2 - 3 248 73
AK299405.1 - 2 - 3 242 73
BC000124.1 - 2 - 3 241 73
BG775086.1 - 1 - 2 111 73

View Full Product Details