Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- CELF2
- Entrez Gene ID
-
10659
- Gene Name
- CUGBP, Elav-like family member 2
- Gene Aliases
-
BRUNOL3, CELF-2, CUG-BP2, CUGBP2, ETR-3, ETR3, NAPOR
- Chromosome Location
-
Chr.10: 10462515 - 11336675 on Build GRCh38
View Full Product Details