The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001129885.1 | NP_001123357.1 | 2 - 3 | 212 | 77 |
XM_011525115.2 | XP_011523417.1 | 2 - 3 | 2110 | 77 | |
GenBank mRNA | BC157870.1 | - | 2 - 3 | 212 | 77 |