The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001025109.1 | NP_001020280.1 | 6 - 7 | 1066 | 101 |
NM_001773.2 | NP_001764.1 | 6 - 7 | 1066 | 101 | |
GenBank mRNA | AF202879.1 | - | 4 - 5 | 311 | 101 |
AF523361.1 | - | 6 - 7 | 879 | 101 | |
AF538601.1 | - | 5 - 6 | 717 | 101 | |
AK091890.1 | - | 3 - 4 | 783 | 101 | |
AK291529.1 | - | 6 - 7 | 819 | 101 | |
AK293030.1 | - | 6 - 7 | 843 | 101 | |
AK294381.1 | - | 5 - 6 | 590 | 101 | |
AK300835.1 | - | 7 - 8 | 883 | 101 | |
BC039146.1 | - | 6 - 7 | 816 | 101 | |
BX538076.1 | - | 3 - 4 | 994 | 101 | |
M81104.1 | - | 6 - 7 | 1065 | 101 | |
S53910.1 | - | 6 - 7 | 898 | 101 | |
S53911.1 | - | 6 - 7 | 898 | 101 |