Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
R3HDM1
Entrez Gene ID
23518
Gene Name
R3H domain containing 1
Gene Aliases
R3HDM
Chromosome Location
Chr.2: 135531429 - 135725270 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001282798.1 NP_001269727.1 6 - 7 857 128
NM_001282799.1 NP_001269728.1 4 - 5 720 128
NM_001282800.1 NP_001269729.1 3 - 4 514 128
NM_015361.3 NP_056176.2 6 - 7 852 128
XM_005263626.3 XP_005263683.1 6 - 7 841 128
XM_005263627.3 XP_005263684.1 6 - 7 841 128
XM_005263628.3 XP_005263685.1 6 - 7 841 128
XM_005263629.3 XP_005263686.1 6 - 7 841 128
XM_006712389.2 XP_006712452.1 6 - 7 841 128
XM_011510870.1 XP_011509172.1 6 - 7 841 128
XM_011510871.1 XP_011509173.1 6 - 7 857 128
XM_011510872.2 XP_011509174.1 5 - 6 799 128
XM_011510873.2 XP_011509175.1 6 - 7 841 128
XM_011510874.2 XP_011509176.1 4 - 5 709 128
XM_011510875.2 XP_011509177.1 4 - 5 725 128
XM_011510876.2 XP_011509178.1 6 - 7 841 128
XM_011510877.2 XP_011509179.1 6 - 7 841 128
XM_011510878.2 XP_011509180.1 6 - 7 841 128
XM_011510879.2 XP_011509181.1 6 - 7 841 128
XM_011510881.1 XP_011509183.1 6 - 7 841 128
XM_017003718.1 XP_016859207.1 6 - 7 857 128
XM_017003719.1 XP_016859208.1 5 - 6 836 128
XM_017003720.1 XP_016859209.1 6 - 7 857 128
XM_017003721.1 XP_016859210.1 6 - 7 857 128
XM_017003722.1 XP_016859211.1 4 - 5 709 128
XM_017003723.1 XP_016859212.1 4 - 5 725 128
XM_017003724.1 XP_016859213.1 5 - 6 799 128
XM_017003725.1 XP_016859214.1 7 - 8 933 128
XM_017003726.1 XP_016859215.1 7 - 8 1003 128
XM_017003727.1 XP_016859216.1 6 - 7 961 128
XM_017003728.1 XP_016859217.1 6 - 7 841 128
XM_017003731.1 XP_016859220.1 7 - 8 954 128
XM_017003732.1 XP_016859221.1 7 - 8 932 128
XM_017003733.1 XP_016859222.1 6 - 7 843 128
XM_017003734.1 XP_016859223.1 7 - 8 1003 128
XM_017003735.1 XP_016859224.1 4 - 5 601 128
GenBank mRNA AK127793.1 - 3 - 4 485 128
AK290015.1 - 6 - 7 743 128
BC041093.1 - 6 - 7 794 128
D21852.1 - 4 - 5 674 128

View Full Product Details