Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
R3HDM1
Entrez Gene ID
23518
Gene Name
R3H domain containing 1
Gene Aliases
R3HDM
Chromosome Location
Chr.2: 135531429 - 135725270 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001282798.1 NP_001269727.1 11 - 12 1342 82
NM_001282799.1 NP_001269728.1 9 - 10 1205 82
NM_001282800.1 NP_001269729.1 8 - 9 999 82
NM_015361.3 NP_056176.2 11 - 12 1337 82
XM_005263626.3 XP_005263683.1 11 - 12 1326 82
XM_005263627.3 XP_005263684.1 11 - 12 1326 82
XM_005263628.3 XP_005263685.1 11 - 12 1326 82
XM_005263629.3 XP_005263686.1 11 - 12 1326 82
XM_006712389.2 XP_006712452.1 11 - 12 1326 82
XM_011510870.1 XP_011509172.1 11 - 12 1326 82
XM_011510871.1 XP_011509173.1 11 - 12 1342 82
XM_011510872.2 XP_011509174.1 10 - 11 1284 82
XM_011510873.2 XP_011509175.1 11 - 12 1326 82
XM_011510874.2 XP_011509176.1 9 - 10 1194 82
XM_011510875.2 XP_011509177.1 9 - 10 1210 82
XM_011510876.2 XP_011509178.1 11 - 12 1326 82
XM_011510877.2 XP_011509179.1 11 - 12 1326 82
XM_011510878.2 XP_011509180.1 11 - 12 1326 82
XM_011510879.2 XP_011509181.1 11 - 12 1326 82
XM_011510881.1 XP_011509183.1 11 - 12 1326 82
XM_017003718.1 XP_016859207.1 11 - 12 1342 82
XM_017003719.1 XP_016859208.1 10 - 11 1321 82
XM_017003720.1 XP_016859209.1 11 - 12 1342 82
XM_017003721.1 XP_016859210.1 11 - 12 1342 82
XM_017003722.1 XP_016859211.1 9 - 10 1194 82
XM_017003723.1 XP_016859212.1 9 - 10 1210 82
XM_017003724.1 XP_016859213.1 10 - 11 1284 82
XM_017003725.1 XP_016859214.1 12 - 13 1418 82
XM_017003726.1 XP_016859215.1 12 - 13 1488 82
XM_017003727.1 XP_016859216.1 11 - 12 1446 82
XM_017003728.1 XP_016859217.1 11 - 12 1326 82
XM_017003729.1 XP_016859218.1 10 - 11 1263 82
XM_017003730.1 XP_016859219.1 8 - 9 1007 82
XM_017003731.1 XP_016859220.1 12 - 13 1439 82
XM_017003732.1 XP_016859221.1 12 - 13 1417 82
XM_017003733.1 XP_016859222.1 12 - 13 1461 82
XM_017003734.1 XP_016859223.1 13 - 14 1621 82
XM_017003735.1 XP_016859224.1 10 - 11 1219 82
XM_017003736.1 XP_016859225.1 11 - 12 1406 82
GenBank mRNA AK127793.1 - 8 - 9 970 82
AK290015.1 - 11 - 12 1228 82
AK297000.1 - 4 - 5 375 82
BC041093.1 - 11 - 12 1279 82
D21852.1 - 9 - 10 1159 82

View Full Product Details