The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_004608.3 | NP_004599.2 | 6 - 7 | 967 | 100 |
XM_005255523.2 | XP_005255580.1 | 7 - 8 | 1252 | 100 | |
XM_011545926.2 | XP_011544228.1 | 7 - 8 | 1415 | 100 | |
XM_017023614.1 | XP_016879103.1 | 7 - 8 | 992 | 100 | |
GenBank mRNA | AJ007989.1 | - | 6 - 7 | 931 | 100 |
BC026031.1 | - | 6 - 7 | 967 | 100 | |
KJ534981.1 | - | 6 - 7 | 1003 | 100 | |
KJ535035.1 | - | 5 - 6 | 1010 | 100 |