Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
TRMT2B
Entrez Gene ID
79979
Gene Name
tRNA methyltransferase 2 homolog B
Gene Aliases
CXorf34, dJ341D10.3
Chromosome Location
Chr.X: 100973358 - 101052118 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001167970.1 NP_001161442.1 6 - 7 951 110
NM_001167971.1 NP_001161443.1 6 - 7 932 110
NM_001167972.1 NP_001161444.1 5 - 6 894 110
NM_024917.5 NP_079193.2 6 - 7 1344 110
XM_005262195.2 XP_005262252.1 7 - 8 1067 110
XM_005262196.2 XP_005262253.1 5 - 6 862 110
XM_005262201.2 XP_005262258.1 3 - 4 442 110
XM_006724705.2 XP_006724768.1 6 - 7 1006 110
XM_011531042.2 XP_011529344.1 3 - 4 560 110
XM_011531044.2 XP_011529346.1 5 - 6 621 110
XM_017029858.1 XP_016885347.1 6 - 7 1339 110
XM_017029859.1 XP_016885348.1 5 - 6 654 110
XM_017029860.1 XP_016885349.1 6 - 7 953 110
XM_017029861.1 XP_016885350.1 5 - 6 863 110
XM_017029862.1 XP_016885351.1 5 - 6 1204 110
XM_017029863.1 XP_016885352.1 7 - 8 876 110
XM_017029864.1 XP_016885353.1 5 - 6 1068 110
XM_017029865.1 XP_016885354.1 5 - 6 696 110
XM_017029866.1 XP_016885355.1 6 - 7 759 110
XM_017029867.1 XP_016885356.1 6 - 7 796 110
XM_017029868.1 XP_016885357.1 5 - 6 669 110
XM_017029869.1 XP_016885358.1 4 - 5 1013 110
XM_017029870.1 XP_016885359.1 4 - 5 603 110
XM_017029871.1 XP_016885360.1 4 - 5 585 110
XM_017029872.1 XP_016885361.1 5 - 6 696 110
XM_017029873.1 XP_016885362.1 5 - 6 671 110
XM_017029874.1 XP_016885363.1 4 - 5 1013 110
XM_017029875.1 XP_016885364.1 3 - 4 560 110
GenBank mRNA AK022749.1 - 5 - 6 707 110
AK315780.1 - 6 - 7 1101 110
AL832849.1 - 6 - 7 870 110
BC007526.1 - 5 - 6 850 110
BC009437.1 - 5 - 6 837 110
BC020116.1 - 6 - 7 905 110
BC034272.2 - 6 - 7 1309 110
BX647969.1 - 5 - 6 891 110

View Full Product Details