Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
TRMT2B
Entrez Gene ID
79979
Gene Name
tRNA methyltransferase 2 homolog B
Gene Aliases
CXorf34, dJ341D10.3
Chromosome Location
Chr.X: 100973358 - 101052118 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001167970.1 NP_001161442.1 7 - 8 1022 93
NM_001167971.1 NP_001161443.1 7 - 8 1003 93
NM_001167972.1 NP_001161444.1 6 - 7 965 93
NM_024917.5 NP_079193.2 7 - 8 1415 93
XM_005262195.2 XP_005262252.1 8 - 9 1138 93
XM_005262196.2 XP_005262253.1 6 - 7 933 93
XM_005262201.2 XP_005262258.1 4 - 5 513 93
XM_006724705.2 XP_006724768.1 7 - 8 1077 93
XM_011531042.2 XP_011529344.1 4 - 5 631 93
XM_011531044.2 XP_011529346.1 6 - 7 692 93
XM_017029858.1 XP_016885347.1 7 - 8 1410 93
XM_017029859.1 XP_016885348.1 6 - 7 725 93
XM_017029860.1 XP_016885349.1 7 - 8 1024 93
XM_017029861.1 XP_016885350.1 6 - 7 934 93
XM_017029862.1 XP_016885351.1 6 - 7 1275 93
XM_017029863.1 XP_016885352.1 8 - 9 947 93
XM_017029864.1 XP_016885353.1 6 - 7 1139 93
XM_017029865.1 XP_016885354.1 6 - 7 767 93
XM_017029866.1 XP_016885355.1 7 - 8 830 93
XM_017029867.1 XP_016885356.1 7 - 8 867 93
XM_017029868.1 XP_016885357.1 6 - 7 740 93
XM_017029869.1 XP_016885358.1 5 - 6 1084 93
XM_017029870.1 XP_016885359.1 5 - 6 674 93
XM_017029871.1 XP_016885360.1 5 - 6 656 93
XM_017029872.1 XP_016885361.1 6 - 7 767 93
XM_017029873.1 XP_016885362.1 6 - 7 742 93
XM_017029874.1 XP_016885363.1 5 - 6 1084 93
XM_017029875.1 XP_016885364.1 4 - 5 631 93
GenBank mRNA AK022749.1 - 6 - 7 778 93
AK315780.1 - 7 - 8 1172 93
AL832849.1 - 7 - 8 941 93
BC007526.1 - 6 - 7 921 93
BC009437.1 - 6 - 7 908 93
BC020116.1 - 7 - 8 976 93
BC034272.2 - 7 - 8 1380 93
BX647969.1 - 6 - 7 962 93

View Full Product Details