The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_014681.5 | NP_055496.2 | 14 - 15 | 3329 | 55 |
XM_005259443.3 | XP_005259500.1 | 14 - 15 | 3840 | 55 | |
XM_011527550.2 | XP_011525852.1 | 14 - 15 | 3964 | 55 | |
XM_011527551.2 | XP_011525853.1 | 14 - 15 | 3278 | 55 | |
XM_017027524.1 | XP_016883013.1 | 13 - 14 | 3243 | 55 | |
GenBank mRNA | AK123046.1 | - | 6 - 7 | 2139 | 55 |
AK297692.1 | - | 9 - 10 | 1488 | 55 | |
AW886612.1 | - | 3 - 4 | 535 | 55 | |
BQ643547.1 | - | 1 - 2 | 47 | 55 | |
BU858266.1 | - | 3 - 4 | 457 | 55 | |
D50924.1 | - | 9 - 10 | 2380 | 55 |