Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
DEC1
Entrez Gene ID
50514
Gene Name
deleted in esophageal cancer 1
Gene Aliases
CTS9
Chromosome Location
Chr.9: 115141818 - 115402644 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_017418.2 NP_059114.1 4 - 5 451 115

View Full Product Details