Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
CCDC90B
Entrez Gene ID
60492
Gene Name
coiled-coil domain containing 90B
Gene Aliases
MDS011, MDS025
Chromosome Location
Chr.11: 83259093 - 83286407 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001286116.1 NP_001273045.1 6 - 7 748 143
NM_001286117.1 NP_001273046.1 6 - 7 1295 143
NM_001286118.1 NP_001273047.1 5 - 6 871 143
NM_001286119.1 NP_001273048.1 6 - 7 956 143
NM_001286120.1 NP_001273049.1 4 - 5 751 143
NM_021825.4 NP_068597.2 6 - 7 975 143
XM_005274154.2 XP_005274211.1 5 - 6 1130 143
XM_005274156.3 XP_005274213.1 6 - 7 962 143
XM_005274159.3 XP_005274216.1 7 - 8 641 143
XM_017018103.1 XP_016873592.1 5 - 6 573 143
XM_017018104.1 XP_016873593.1 7 - 8 1066 143
GenBank mRNA AF182412.1 - 6 - 7 545 143
AF182424.1 - 6 - 7 545 143
AF271782.1 - 6 - 7 902 143
AK055972.1 - 6 - 7 1292 143
AK225726.1 - 6 - 7 556 143
AK304769.1 - 6 - 7 708 143
AK315787.1 - 6 - 7 936 143
AL136791.1 - 6 - 7 695 143
AV756714.1 - 5 - 6 511 143
BC014573.1 - 6 - 7 585 143
BC020783.1 - 6 - 7 546 143
BC067764.1 - 6 - 7 866 143
BC107755.1 - 6 - 7 716 143
BG700162.1 - 4 - 5 328 144

View Full Product Details