Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ATP5G1
Entrez Gene ID
516
Gene Name
ATP synthase, H+ transporting, mitochondrial Fo complex subunit C1 (subunit 9)
Gene Aliases
ATP5A, ATP5G
Chromosome Location
Chr.17: 48892786 - 48895871 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_005175.2 NP_005166.1 1 - 2 119 103

View Full Product Details