The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_022336.3 | NP_071731.1 | 11 - 12 | 1456 | 107 |
XM_006712204.1 | XP_006712267.1 | 10 - 11 | 1436 | 107 | |
XM_011510502.2 | XP_011508804.2 | 10 - 11 | 1315 | 107 | |
XM_011510503.2 | XP_011508805.2 | 11 - 12 | 1212 | 107 | |
GenBank mRNA | AF130988.1 | - | 11 - 12 | 1457 | 107 |
AK296936.1 | - | 10 - 11 | 1346 | 107 | |
AK313781.1 | - | 11 - 12 | 1340 | 107 | |
BC093870.1 | - | 10 - 11 | 1048 | 107 | |
BC093872.1 | - | 10 - 11 | 1048 | 107 | |
CR749352.1 | - | 11 - 12 | 1565 | 107 |