The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001199198.2 | NP_001186127.1 | 10 - 11 | 1291 | 69 |
NM_018309.4 | NP_060779.2 | 10 - 11 | 1291 | 69 | |
XM_011512974.2 | XP_011511276.1 | 10 - 11 | 1154 | 69 | |
XM_017006841.1 | XP_016862330.1 | 10 - 11 | 1122 | 69 | |
GenBank mRNA | AB449910.1 | - | 10 - 11 | 1089 | 69 |
AK001750.1 | - | 7 - 8 | 777 | 69 | |
AL713684.1 | - | 2 - 3 | 172 | 69 | |
BC020955.1 | - | 10 - 11 | 1085 | 69 |