Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ADAM32
Entrez Gene ID
203102
Gene Name
ADAM metallopeptidase domain 32
Gene Aliases
-
Chromosome Location
Chr.8: 39107529 - 39284917 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_145004.6 NP_659441.3 12 - 13 1483 99
XM_006716298.2 XP_006716361.1 12 - 13 1277 99
XM_011544433.1 XP_011542735.1 12 - 13 1277 99
XM_011544434.1 XP_011542736.1 10 - 11 1058 99
XM_017013183.1 XP_016868672.1 11 - 12 1208 99
XM_017013184.1 XP_016868673.1 12 - 13 1277 99
XM_017013185.1 XP_016868674.1 11 - 12 1283 99
XM_017013186.1 XP_016868675.1 8 - 9 887 99
XM_017013187.1 XP_016868676.1 4 - 5 423 99
XM_017013188.1 XP_016868677.1 3 - 4 392 99
GenBank mRNA AK131038.1 - 4 - 5 452 99
AK313864.1 - 12 - 13 1304 99
AY358739.1 - 12 - 13 1363 99
BC026085.2 - 12 - 13 1267 99
BC026169.2 - 12 - 13 1294 99
BC028702.2 - 12 - 13 1277 99
BC030014.2 - 12 - 13 1270 99
BC030698.2 - 12 - 13 1285 99
BC034975.2 - 12 - 13 1278 99

View Full Product Details