The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001313994.1 | NP_001300923.1 | 11 - 12 | 1368 | 64 |
NM_145004.6 | NP_659441.3 | 14 - 15 | 1766 | 64 | |
XM_006716298.2 | XP_006716361.1 | 14 - 15 | 1560 | 64 | |
XM_006716299.1 | XP_006716362.1 | 12 - 13 | 1242 | 64 | |
XM_011544433.1 | XP_011542735.1 | 14 - 15 | 1560 | 64 | |
XM_011544434.1 | XP_011542736.1 | 12 - 13 | 1341 | 64 | |
XM_017013183.1 | XP_016868672.1 | 13 - 14 | 1491 | 64 | |
XM_017013184.1 | XP_016868673.1 | 14 - 15 | 1560 | 64 | |
XM_017013185.1 | XP_016868674.1 | 13 - 14 | 1566 | 64 | |
XM_017013186.1 | XP_016868675.1 | 10 - 11 | 1170 | 64 | |
XM_017013187.1 | XP_016868676.1 | 6 - 7 | 706 | 64 | |
XM_017013188.1 | XP_016868677.1 | 5 - 6 | 675 | 64 | |
GenBank mRNA | AK122608.1 | - | 12 - 13 | 1121 | 64 |
AK131038.1 | - | 6 - 7 | 735 | 64 | |
AK300148.1 | - | 11 - 12 | 1326 | 64 | |
AK313864.1 | - | 14 - 15 | 1587 | 64 | |
AY172952.1 | - | 12 - 13 | 1370 | 64 | |
AY358739.1 | - | 14 - 15 | 1646 | 64 | |
BC026085.2 | - | 14 - 15 | 1550 | 64 | |
BC026169.2 | - | 14 - 15 | 1577 | 64 | |
BC028702.2 | - | 14 - 15 | 1560 | 64 | |
BC030014.2 | - | 14 - 15 | 1553 | 64 | |
BC030698.2 | - | 14 - 15 | 1568 | 64 | |
BC034975.2 | - | 14 - 15 | 1561 | 64 |