Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ADAM32
Entrez Gene ID
203102
Gene Name
ADAM metallopeptidase domain 32
Gene Aliases
-
Chromosome Location
Chr.8: 39107529 - 39284917 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001313994.1 NP_001300923.1 5 - 6 697 134
NM_145004.6 NP_659441.3 6 - 7 777 134
XM_006716298.2 XP_006716361.1 6 - 7 571 134
XM_006716299.1 XP_006716362.1 6 - 7 571 134
XM_011544433.1 XP_011542735.1 6 - 7 571 134
XM_011544434.1 XP_011542736.1 6 - 7 571 134
XM_017013184.1 XP_016868673.1 6 - 7 571 134
XM_017013185.1 XP_016868674.1 5 - 6 577 134
GenBank mRNA AB209690.1 - 6 - 7 558 134
AK122608.1 - 6 - 7 571 134
AK300148.1 - 5 - 6 655 134
AK313864.1 - 6 - 7 598 134
AY172952.1 - 6 - 7 699 134
AY358739.1 - 6 - 7 657 134
BC026085.2 - 6 - 7 561 134
BC026169.2 - 6 - 7 588 134
BC028702.2 - 6 - 7 571 134
BC030014.2 - 6 - 7 564 134
BC030698.2 - 6 - 7 579 134
BC034975.2 - 6 - 7 572 134

View Full Product Details