Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CHDH
Entrez Gene ID
55349
Gene Name
choline dehydrogenase
Gene Aliases
-
Chromosome Location
Chr.3: 53813130 - 53846393 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_018397.4 NP_060867.2 8 - 9 1802 63
XM_006713250.3 XP_006713313.1 8 - 9 1584 63
XM_006713251.3 XP_006713314.1 8 - 9 1761 63
XM_006713252.3 XP_006713315.1 7 - 8 1441 63
XM_011533938.2 XP_011532240.1 8 - 9 1618 63
XM_011533939.2 XP_011532241.1 8 - 9 2457 63
XM_017006797.1 XP_016862286.1 8 - 9 1615 63
XM_017006798.1 XP_016862287.1 8 - 9 1607 63
XM_017006799.1 XP_016862288.1 7 - 8 1438 63
GenBank mRNA AJ272267.1 - 6 - 7 1026 63
AK297575.1 - 4 - 5 485 63
BC034502.1 - 8 - 9 1560 63

View Full Product Details