Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
RC3H2
Entrez Gene ID
54542
Gene Name
ring finger and CCCH-type domains 2
Gene Aliases
MNAB, RNF164
Chromosome Location
Chr.9: 122849453 - 122905283 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001100588.1 NP_001094058.1 11 - 12 2088 60
NM_018835.2 NP_061323.2 11 - 12 2088 60
GenBank mRNA AF255303.1 - 10 - 11 2448 60
AK000308.1 - 9 - 10 1522 60
AK027042.1 - 2 - 3 325 60
AK226010.1 - 9 - 10 1522 60
AL833177.1 - 12 - 13 1945 60
AL834431.1 - 5 - 6 839 60

View Full Product Details