The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001100588.1 | NP_001094058.1 | 15 - 16 | 3050 | 86 |
NM_018835.2 | NP_061323.2 | 15 - 16 | 3050 | 86 | |
GenBank mRNA | AF255303.1 | - | 14 - 15 | 3410 | 86 |
AF255304.1 | - | 3 - 4 | 494 | 86 | |
AK000308.1 | - | 13 - 14 | 2484 | 86 | |
AK000720.1 | - | 2 - 3 | 277 | 86 | |
AK027042.1 | - | 6 - 7 | 1287 | 86 | |
AK226010.1 | - | 13 - 14 | 2484 | 86 | |
AL833177.1 | - | 16 - 17 | 2907 | 86 | |
AL834431.1 | - | 9 - 10 | 1801 | 86 |