Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
PHF7
Entrez Gene ID
51533
Gene Name
PHD finger protein 7
Gene Aliases
HSPC045, HSPC226, NYD-SP6
Chromosome Location
Chr.3: 52410508 - 52423641 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001278221.2 NP_001265150.1 3 - 4 903 59
NM_001321126.1 NP_001308055.1 3 - 4 862 59
NM_001321127.1 NP_001308056.1 3 - 4 995 59
NM_016483.6 NP_057567.3 3 - 4 903 59
XM_011533826.2 XP_011532128.1 3 - 4 853 59
GenBank mRNA AK223531.1 - 2 - 3 120 59
AK292221.1 - 3 - 4 709 59
AY014283.2 - 3 - 4 477 59
BC022002.1 - 3 - 4 762 59
BQ669125.1 - 3 - 4 613 59

View Full Product Details