Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
PHF7
Entrez Gene ID
51533
Gene Name
PHD finger protein 7
Gene Aliases
HSPC045, HSPC226, NYD-SP6
Chromosome Location
Chr.3: 52410508 - 52423641 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001278221.2 NP_001265150.1 4 - 5 991 115
NM_001321126.1 NP_001308055.1 4 - 5 950 115
NM_001321127.1 NP_001308056.1 4 - 5 1083 115
NM_016483.6 NP_057567.3 4 - 5 991 115
XM_011533826.2 XP_011532128.1 4 - 5 941 115
GenBank mRNA AK223531.1 - 3 - 4 208 115
AK292221.1 - 4 - 5 797 115
AY014283.2 - 4 - 5 565 115
BC022002.1 - 4 - 5 850 115

View Full Product Details