Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
GTPBP3
Entrez Gene ID
84705
Gene Name
GTP binding protein 3 (mitochondrial)
Gene Aliases
COXPD23, GTPBG3, MSS1, MTGP1, THDF1
Chromosome Location
Chr.19: 17334982 - 17342731 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001128855.2 NP_001122327.1 5 - 6 758 68
NM_001195422.1 NP_001182351.1 5 - 6 751 68
NM_032620.3 NP_116009.2 5 - 6 758 68
GenBank mRNA AF360742.1 - 5 - 6 720 68
AF360743.1 - 5 - 6 720 68
AF360744.1 - 5 - 6 700 68
AK027606.1 - 5 - 6 705 68
AK291929.1 - 5 - 6 702 68
AK297953.1 - 5 - 6 733 68
AK298485.1 - - 981 68
AK311442.1 - 5 - 6 821 68
AY078988.1 - 5 - 6 720 68
BC008924.2 - 3 - 4 298 68
BC017207.2 - 5 - 6 690 68
BC019261.1 - 5 - 6 699 68
BM789389.1 - 2 - 3 222 68

View Full Product Details