Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SUN2
Entrez Gene ID
25777
Gene Name
Sad1 and UNC84 domain containing 2
Gene Aliases
UNC84B
Chromosome Location
Chr.22: 38734714 - 38756019 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001199579.1 NP_001186508.1 6 - 7 977 63
NM_001199580.1 NP_001186509.1 7 - 8 913 63
NM_015374.2 NP_056189.1 6 - 7 914 63
XM_011530104.2 XP_011528406.1 5 - 6 771 63
XM_011530105.2 XP_011528407.1 6 - 7 638 63
XM_017028748.1 XP_016884237.1 6 - 7 909 63
GenBank mRNA AB014568.1 - 6 - 7 696 63
AK295787.1 - 6 - 7 638 63
AK304188.1 - 7 - 8 773 63
AY682988.1 - 6 - 7 695 63
BC094797.1 - 6 - 7 934 63
BC111549.1 - 5 - 6 643 63
BC111717.1 - 5 - 6 643 63
BX537962.1 - 7 - 8 905 63
CR456474.1 - 5 - 6 641 63

View Full Product Details