The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001301075.1 | NP_001288004.1 | 1 - 2 | 303 | 104 |
NM_004779.5 | NP_004770.4 | 1 - 2 | 303 | 104 | |
XM_005268527.2 | XP_005268584.1 | 2 - 3 | 757 | 104 | |
XM_017010049.1 | XP_016865538.1 | 2 - 3 | 757 | 104 | |
GenBank mRNA | AF053318.1 | - | 1 - 2 | 177 | 104 |
AK297381.1 | - | 1 - 2 | 168 | 104 | |
AL122045.1 | - | 1 - 2 | 184 | 104 |