Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ARMCX3
Entrez Gene ID
51566
Gene Name
armadillo repeat containing, X-linked 3
Gene Aliases
ALEX3, GASP6, dJ545K15.2
Chromosome Location
Chr.X: 101623127 - 101631910 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_177947.2 NP_808816.1 1 - 2 147 70
GenBank mRNA AL708980.1 - 1 - 2 146 70
BC005194.1 - 1 - 2 91 70
BG035798.1 - 1 - 2 160 70

View Full Product Details