The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001317946.1 | NP_001304875.1 | 4 - 5 | 761 | 65 |
XM_011530345.2 | XP_011528647.1 | 4 - 5 | 757 | 65 | |
GenBank mRNA | AK024025.1 | - | 4 - 5 | 729 | 65 |
CB161660.1 | - | 2 - 3 | 207 | 65 |