Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
PCDHB4
Entrez Gene ID
56131
Gene Name
protocadherin beta 4
Gene Aliases
PCDH-BETA4
Chromosome Location
Chr.5: 141121750 - 141125623 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_018938.3 NP_061761.1 1 - 1 3621 129
GenBank mRNA AF217754.1 - 1 - 1 3372 129
AW135510.1 - 1 - 1 129

View Full Product Details