Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
SPATA20
Entrez Gene ID
64847
Gene Name
spermatogenesis associated 20
Gene Aliases
HEL-S-98, SSP411, Tisp78
Chromosome Location
Chr.17: 50547089 - 50555852 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001258373.1 NP_001245302.1 2 - 3 299 192
NM_022827.3 NP_073738.2 2 - 3 244 137
GenBank mRNA AK092731.1 - 2 - 3 229 192
AK125807.1 - 4 - 5 974 192
AK291805.1 - 2 - 3 187 192
BC025255.1 - 2 - 3 132 137
BC065526.1 - 2 - 3 186 192
DA295930.1 - 2 - 3 259 192
DA489234.1 - 2 - 3 244 137

View Full Product Details