Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
SPATA20
Entrez Gene ID
64847
Gene Name
spermatogenesis associated 20
Gene Aliases
HEL-S-98, SSP411, Tisp78
Chromosome Location
Chr.17: 50547089 - 50555852 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001258372.1 NP_001245301.1 4 - 5 582 82
NM_001258373.1 NP_001245302.1 5 - 6 685 82
NM_022827.3 NP_073738.2 5 - 6 630 82
GenBank mRNA AK025000.1 - 4 - 5 479 82
AK092731.1 - 5 - 6 615 82
AK125807.1 - 7 - 8 1360 82
AK225189.1 - 4 - 5 479 82
AK291805.1 - 5 - 6 573 82
AK308616.1 - 5 - 6 755 82
AK310416.1 - 4 - 5 474 82
BC025255.1 - 5 - 6 518 82
BC065526.1 - 5 - 6 572 82
BC111029.1 - 4 - 5 487 82
BM787432.1 - 4 - 5 545 82
CR936769.1 - 4 - 5 500 82
DQ238597.1 - 4 - 5 548 82
GQ472220.1 - 4 - 5 487 82

View Full Product Details