Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
MKLN1
Entrez Gene ID
4289
Gene Name
muskelin 1
Gene Aliases
TWA2
Chromosome Location
Chr.7: 131110096 - 131496639 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001145354.1 NP_001138826.1 17 - 18 2168 93
NM_001321316.1 NP_001308245.1 16 - 17 2099 93
NM_013255.4 NP_037387.2 16 - 17 2096 93
GenBank mRNA AF047489.1 - 16 - 17 2050 93
AK002024.1 - 16 - 17 2038 93
AK294384.1 - 17 - 18 2169 93
AL162071.1 - 7 - 8 1076 93
BC002834.2 - 11 - 12 1474 93
BC067825.1 - 16 - 17 2053 93

View Full Product Details