The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001145354.1 | NP_001138826.1 | 4 - 5 | 450 | 123 |
NM_001321316.1 | NP_001308245.1 | 3 - 4 | 378 | 123 | |
NM_013255.4 | NP_037387.2 | 3 - 4 | 378 | 123 | |
XM_006715993.2 | XP_006716056.1 | 3 - 4 | 378 | 123 | |
XM_011516224.2 | XP_011514526.1 | 3 - 4 | 378 | 123 | |
GenBank mRNA | AF047489.1 | - | 3 - 4 | 332 | 123 |
AK002024.1 | - | 3 - 4 | 317 | 123 | |
AK294384.1 | - | 4 - 5 | 451 | 123 | |
AK310452.1 | - | 3 - 4 | 331 | 123 | |
AL705061.1 | - | 3 - 4 | 322 | 123 | |
BC067825.1 | - | 3 - 4 | 335 | 123 | |
BU945366.1 | - | - | 339 | 123 | |
DA172589.1 | - | 3 - 4 | 378 | 123 |