The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001204366.1 | NP_001191295.1 | 1 - 2 | 310 | 135 |
NM_001204368.1 | NP_001191297.1 | 1 - 2 | 310 | 135 | |
NM_002413.4 | NP_002404.1 | 1 - 2 | 310 | 135 | |
XM_017008211.1 | XP_016863700.1 | 1 - 2 | 322 | 135 | |
XM_017008212.1 | XP_016863701.1 | 1 - 2 | 322 | 135 | |
XM_017008213.1 | XP_016863702.1 | 1 - 2 | 322 | 135 | |
GenBank mRNA | AA122237.1 | - | 1 - 2 | 310 | 135 |
AK130948.1 | - | 1 - 2 | 259 | 135 | |
BC025416.1 | - | 1 - 2 | 224 | 135 | |
BE312976.1 | - | 1 - 2 | 274 | 135 | |
CR407640.1 | - | 1 - 2 | 58 | 135 | |
DB257575.1 | - | 1 - 2 | 73 | 136 | |
U77604.1 | - | 1 - 2 | 69 | 135 |