The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001286675.1 | NP_001273604.1 | 1 - 2 | 255 | 62 |
GenBank mRNA | AF366055.1 | - | 1 - 2 | 163 | 62 |