The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001142857.1 | NP_001136329.1 | 1 - 2 | 223 | 86 |
NR_024605.1 | 1 - 2 | 223 | 86 | ||
XM_017013826.1 | XP_016869315.1 | 1 - 2 | 237 | 86 | |
GenBank mRNA | AK307101.1 | - | 1 - 2 | 213 | 86 |
AK316037.1 | - | 1 - 2 | 177 | 86 | |
BP375493.1 | - | 1 - 2 | 172 | 86 | |
DA174619.1 | - | 1 - 2 | 173 | 86 |