The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001301227.1 | NP_001288156.1 | 6 - 7 | 873 | 82 |
NM_213674.1 | NP_998839.1 | 6 - 7 | 873 | 82 | |
XM_017015090.1 | XP_016870579.1 | 6 - 7 | 745 | 82 | |
GenBank mRNA | AI360640.1 | - | 3 - 4 | 82 | |
AK130045.1 | - | 6 - 7 | 461 | 82 | |
AK223258.1 | - | 6 - 7 | 742 | 82 | |
BC011776.2 | - | 6 - 7 | 693 | 82 | |
EU106177.1 | - | 6 - 7 | 634 | 82 | |
FJ224304.1 | - | 6 - 7 | 873 | 82 | |
M12125.1 | - | 6 - 7 | 752 | 82 | |
M74817.1 | - | 6 - 7 | 723 | 82 | |
M75165.1 | - | 6 - 7 | 723 | 82 |