Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
LRRFIP2
Entrez Gene ID
9209
Gene Name
LRR binding FLII interacting protein 2
Gene Aliases
HUFI-2
Chromosome Location
Chr.3: 37052626 - 37176817 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_006309.3 NP_006300.1 10 - 11 683 67
XM_005265539.2 XP_005265596.1 11 - 12 1683 67
XM_005265540.1 XP_005265597.1 12 - 13 966 67
XM_006713385.1 XP_006713448.1 11 - 12 948 67
XM_006713386.1 XP_006713449.1 10 - 11 909 67
XM_006713389.1 XP_006713452.1 10 - 11 900 67
XM_006713390.1 XP_006713453.1 11 - 12 943 67
XM_006713392.1 XP_006713455.1 11 - 12 951 67
XM_006713393.1 XP_006713456.1 11 - 12 951 67
XM_006713394.1 XP_006713457.1 9 - 10 858 67
XM_006713395.1 XP_006713458.1 11 - 12 951 67
XM_006713396.1 XP_006713459.1 9 - 10 843 67
XM_006713397.1 XP_006713460.1 11 - 12 951 67
XM_011534217.1 XP_011532519.1 11 - 12 951 67
XM_011534218.1 XP_011532520.1 12 - 13 994 67
XM_011534219.1 XP_011532521.1 12 - 13 1062 67
XM_011534221.1 XP_011532523.1 9 - 10 843 67
XM_011534222.1 XP_011532524.1 11 - 12 951 67
XM_017007465.1 XP_016862954.1 8 - 9 798 67
XM_017007466.1 XP_016862955.1 11 - 12 951 67
XM_017007467.1 XP_016862956.1 11 - 12 951 67
XM_017007468.1 XP_016862957.1 6 - 7 690 67
XM_017007469.1 XP_016862958.1 11 - 12 951 67
GenBank mRNA AF115509.1 - 10 - 11 642 67

View Full Product Details