Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
LRRFIP2
Entrez Gene ID
9209
Gene Name
LRR binding FLII interacting protein 2
Gene Aliases
HUFI-2
Chromosome Location
Chr.3: 37052626 - 37176817 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_006309.3 NP_006300.1 11 - 12 735 84
XM_005265539.2 XP_005265596.1 12 - 13 1735 84
XM_005265540.1 XP_005265597.1 13 - 14 1018 84
XM_006713385.1 XP_006713448.1 12 - 13 1000 84
XM_006713386.1 XP_006713449.1 11 - 12 961 84
XM_006713389.1 XP_006713452.1 11 - 12 952 84
XM_006713390.1 XP_006713453.1 12 - 13 995 84
XM_006713392.1 XP_006713455.1 12 - 13 1003 84
XM_006713393.1 XP_006713456.1 12 - 13 1003 84
XM_006713394.1 XP_006713457.1 10 - 11 910 84
XM_006713395.1 XP_006713458.1 12 - 13 1003 84
XM_006713396.1 XP_006713459.1 10 - 11 895 84
XM_011534217.1 XP_011532519.1 12 - 13 1003 84
XM_011534218.1 XP_011532520.1 13 - 14 1046 84
XM_011534219.1 XP_011532521.1 13 - 14 1114 84
XM_011534221.1 XP_011532523.1 10 - 11 895 84
XM_011534222.1 XP_011532524.1 12 - 13 1003 84
XM_017007465.1 XP_016862954.1 9 - 10 850 84
XM_017007466.1 XP_016862955.1 12 - 13 1003 84
XM_017007467.1 XP_016862956.1 12 - 13 1003 84
XM_017007468.1 XP_016862957.1 7 - 8 742 84
XM_017007469.1 XP_016862958.1 12 - 13 1003 84
GenBank mRNA AF115509.1 - 11 - 12 694 84

View Full Product Details