Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
LRRFIP2
Entrez Gene ID
9209
Gene Name
LRR binding FLII interacting protein 2
Gene Aliases
HUFI-2
Chromosome Location
Chr.3: 37052626 - 37176817 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001134369.2 NP_001127841.1 14 - 15 1591 104
NM_001282691.1 NP_001269620.1 18 - 19 1580 104
NM_006309.3 NP_006300.1 27 - 28 2181 104
NM_017724.2 NP_060194.1 13 - 14 1519 104
XM_005265539.2 XP_005265596.1 28 - 29 3181 104
XM_005265540.1 XP_005265597.1 29 - 30 2464 104
XM_005265550.1 XP_005265607.1 20 - 21 2017 104
XM_005265551.1 XP_005265608.1 19 - 20 1906 104
XM_005265553.1 XP_005265610.1 16 - 17 1753 104
XM_005265554.2 XP_005265611.1 16 - 17 2485 104
XM_005265556.1 XP_005265613.1 15 - 16 1681 104
XM_006713385.1 XP_006713448.1 28 - 29 2446 104
XM_006713386.1 XP_006713449.1 27 - 28 2407 104
XM_006713387.1 XP_006713450.1 27 - 28 2404 104
XM_006713388.1 XP_006713451.1 27 - 28 2401 104
XM_006713389.1 XP_006713452.1 27 - 28 2398 104
XM_006713390.1 XP_006713453.1 28 - 29 2441 104
XM_006713392.1 XP_006713455.1 27 - 28 2377 104
XM_006713393.1 XP_006713456.1 27 - 28 2359 104
XM_006713394.1 XP_006713457.1 26 - 27 2356 104
XM_006713395.1 XP_006713458.1 27 - 28 2356 104
XM_006713396.1 XP_006713459.1 26 - 27 2341 104
XM_006713397.1 XP_006713460.1 25 - 26 2233 104
XM_006713400.1 XP_006713463.1 17 - 18 1798 104
XM_006713404.1 XP_006713467.1 14 - 15 1531 104
XM_011534217.1 XP_011532519.1 28 - 29 2449 104
XM_011534218.1 XP_011532520.1 29 - 30 2492 104
XM_011534219.1 XP_011532521.1 29 - 30 2560 104
XM_011534221.1 XP_011532523.1 26 - 27 2341 104
XM_011534222.1 XP_011532524.1 26 - 27 2254 104
XM_011534229.1 XP_011532531.1 18 - 19 1841 104
XM_017007465.1 XP_016862954.1 25 - 26 2296 104
XM_017007466.1 XP_016862955.1 26 - 27 2290 104
XM_017007467.1 XP_016862956.1 26 - 27 2284 104
XM_017007468.1 XP_016862957.1 23 - 24 2188 104
XM_017007469.1 XP_016862958.1 25 - 26 2182 104
XM_017007470.1 XP_016862959.1 23 - 24 2146 104
XM_017007471.1 XP_016862960.1 18 - 19 1855 104
XM_017007472.1 XP_016862961.1 18 - 19 1813 104
XM_017007473.1 XP_016862962.1 17 - 18 1783 104
XM_017007474.1 XP_016862963.1 17 - 18 1762 104
XM_017007475.1 XP_016862964.1 17 - 18 2494 104
XM_017007476.1 XP_016862965.1 17 - 18 1796 104
XM_017007477.1 XP_016862966.1 16 - 17 2422 104
XM_017007478.1 XP_016862967.1 15 - 16 1660 104
XM_017007479.1 XP_016862968.1 16 - 17 1660 104
XM_017007480.1 XP_016862969.1 15 - 16 1904 104
XM_017007481.1 XP_016862970.1 15 - 16 1651 104
XM_017007482.1 XP_016862971.1 14 - 15 1588 104
XM_017007483.1 XP_016862972.1 14 - 15 2290 104
XM_017007484.1 XP_016862973.1 15 - 16 1601 104
GenBank mRNA AF115509.1 - 27 - 28 2140 104
AK000255.1 - 12 - 13 1204 104
AK024692.1 - 6 - 7 537 104
AK124432.1 - 5 - 6 3315 104
AK291514.1 - 14 - 15 1568 104
AK302282.1 - 18 - 19 1554 104
BC053668.1 - 13 - 14 1476 104
BX648794.1 - 10 - 11 970 104
CR749705.1 - 4 - 5 1908 104

View Full Product Details