The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001017520.1 | NP_001017520.1 | 1 - 2 | 374 | 82 |
NM_004088.3 | NP_004079.3 | 1 - 2 | 374 | 82 | |
GenBank mRNA | AB046378.1 | - | 1 - 2 | 306 | 82 |
AK223317.1 | - | 1 - 2 | 218 | 82 | |
AK313615.1 | - | 1 - 2 | 283 | 82 | |
BC012920.1 | - | 1 - 2 | 247 | 82 |